Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D4-abiraterone | 154229-21-7 | 99.85% | 347.50 | 25 MG
SDP

Catalog No. 5000347356
Encompass_Preferred

D4-abiraterone is a significant metabolite of abiraterone. It functions as an inhibitor of CYP17A1, 3β-hydroxysteroid dehydrogenase (3βHSD), and steroid-5a-reductase (SRD5A), and also acts as an antagonist of the androgen receptor.

  • Higher potency than abiraterone in blocking conversion of dehydroepiandrosterone (DHEA) in xenografts.
  • Demonstrates greater affinity for mutant and wild-type androgen receptor compared to abiraterone.
  • Effectively suppresses gene expression in cell lines.
  • Delays tumor progression more significantly than abiraterone acetate.
  • Purity: 99.85%.
  • Molecular Weight: 347.50.
  • Appearance: Solid, white to off-white.
  • Solubility in DMSO: 10 mg/mL (28.78 mM).

Catalog No. 50-003-47356 Supplier Medchemexpress LLC Supplier No. 25MGHY109619
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D4-abiraterone is a significant metabolite of abiraterone. It functions as an inhibitor of CYP17A1, 3β-hydroxysteroid dehydrogenase (3βHSD), and steroid-5a-reductase (SRD5A), and also acts as an antagonist of the androgen receptor.

  • Higher potency than abiraterone in blocking conversion of dehydroepiandrosterone (DHEA) in xenografts.
  • Demonstrates greater affinity for mutant and wild-type androgen receptor compared to abiraterone.
  • Effectively suppresses gene expression in cell lines.
  • Delays tumor progression more significantly than abiraterone acetate.
  • Purity: 99.85%.
  • Molecular Weight: 347.50.
  • Appearance: Solid, white to off-white.
  • Solubility in DMSO: 10 mg/mL (28.78 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.