Learn More
Medchemexpress LLC HY-109619 5mg Medchemexpress, D4-abiraterone CAS:154229-21-7 Purity:>98%
SDP
Catalog No. 501968714
Encompass_Preferred
Medchemexpress, HY-109619 5mg D4-abiraterone CAS:154229-21-7 D4-abiraterone is a major metabolite of abiraterone. D4-abiraterone is an inhibitor of CYP17A1, 3b-hydroxysteroid dehydrogenase (3βHSD) and steroid-5a-reductase (SRD5A) and also an antagonist of androgen receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8714 Supplier Medchemexpress LLC Supplier No. HY1096195MG
Medchemexpress, HY-109619 5mg D4-abiraterone CAS:154229-21-7 D4-abiraterone is a major metabolite of abiraterone. D4-abiraterone is an inhibitor of CYP17A1, 3b-hydroxysteroid dehydrogenase (3βHSD) and steroid-5a-reductase (SRD5A) and also an antagonist of androgen receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.