Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D4 Cyclomethicone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019453 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9453 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-9453 Supplier MilliporeSigma Supelco Supplier No. PHR1565500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 556-67-2
Vapor Density 1 (vs. air)
Density 0.956 g/mL (at 25°C)
Physical Form Liquid
Linear Formula [-Si(CH3)2 O-]4
MDL Number MFCD00003269
Melting Point 17°C to 18°C
Boiling Point 175°C to 176°C
Synonym Octamethylcyclotetrasiloxane
Type Certified Reference Material
Quantity 500 mg
Formula Weight 296.62
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.