Learn More
Biolegend DyLight™ 405 Goat anti-mouse IgG1
Supplier:  Biolegend 409109
Encompass_Preferred
DyLight™ 405 Goat anti-mouse IgG1 [Poly24091]; Isotype: Goat Polyclonal Ig; Reactivity: Mouse; Apps: FC, ICC; Size: 100 ug
Catalog No. 50-169-251