Learn More
Medchemexpress LLC HY-10324 10mg , D4476 CAS:301836-43-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1032410MG
Encompass_Preferred
HY-10324 10mg D4476 CAS: 301836-43-1 D4476 is a potent, selective and cell-permeable inhibitor of casein kinase 1(CK1) with an IC50 value of 0.3 μM in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0443