Learn More
Abcam D4476, CK1 inhibitor, 10MG
Catalog No. 502707781
Encompass_Preferred
MW 398.4 Da, Purity >99%. Selective cell-permeable inhibitor of casein kinase 1 (CK1) . (IC₅₀ values are 0.3 (CK1), 0.5 (ALK5), 9.1 (PKd1) and 5.8 μM (p38αMAPK)). Displays selectivity over many other protein kinases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7781 Supplier Abcam Supplier No. AB12022010MG
MW 398.4 Da, Purity >99%. Selective cell-permeable inhibitor of casein kinase 1 (CK1) . (IC₅₀ values are 0.3 (CK1), 0.5 (ALK5), 9.1 (PKd1) and 5.8 μM (p38αMAPK)). Displays selectivity over many other protein kinases.
The product is subject to the following: Abcam Restricted Use Statement