Learn More
Aviva Systems Biology Corp D4S234E Antibody - N-terminal region (ARP44912_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44912_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against D4S234E. It was validated on Western Blot. D4S234E,DNA segment on chromosome 4 (unique) 234 expressed sequence,P21; NSG1; D4S234; NEEP21,neuron-specific protein family member 1,neuron-specific protein family member 1,OTTHUMP00000155127,OTTHUMP00000159148,OTTHUMP00000217303,OTTHUMP00000217304,OTTHUMP00000217305,brain neuron cytoplasmic protein 1,carboxyterminally EE-tagged neuron-enriched endosomal 21 kDa proteinThe immunogen is a synthetic peptide directed towards the N terminal region of human D4S234E. Alternate names: D4S234, NEEP21, NSG1, P21, D4S234EApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-168