Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies D5000 Reagents, 100 to 5000 bp, 105 Samples

Catalog No. NC1940389
Encompass

The D5000 reagents are designed for the analysis of double-stranded DNA molecules ranging from 100 to 5000 bp. This assay is optimized for use with the Agilent TapeStation system and contains SYBR™ Green I dye for sensitive detection.

  • Quantitative range: 0.1 to 50 ng/μL
  • Sizing range: 100 to 5000 bp
  • Sample volume: 1 μL
  • Kit size: 105 samples
  • Quantitative accuracy: 20 %
  • Sizing accuracy: 10 %
  • For research use only

Catalog No. NC1940389 Supplier Agilent Technologies Supplier No. 50675589
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The D5000 reagents are designed for the analysis of double-stranded DNA molecules ranging from 100 to 5000 bp. This assay is optimized for use with the Agilent TapeStation system and contains SYBR™ Green I dye for sensitive detection.

  • Quantitative range: 0.1 to 50 ng/μL
  • Sizing range: 100 to 5000 bp
  • Sample volume: 1 μL
  • Kit size: 105 samples
  • Quantitative accuracy: 20 %
  • Sizing accuracy: 10 %
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.