Learn More
Agilent Technologies D5000 ScreenTape, DNA Electrophoresis, 100-5000 bp, 7/Pk.
Catalog No. NC1940388
Encompass
The D5000 ScreenTape is designed for the automated electrophoresis of DNA samples, facilitating objective quantification, sizing, and purity determination. This system is essential for quality control in next-generation sequencing (NGS) workflows, as well as for the analysis of PCR and qPCR products. Compatible with all TapeStation instruments, the system utilizes individual lanes and loading tips to prevent cross-contamination and sample carryover.
  • Provides reliable analysis of DNA fragments and smears from 100 bp to 5000 bp.
  • Includes 7 ScreenTape devices, providing a total capacity of 105 samples.
  • Requires a 1 μL sample volume per analysis.
  • Features rapid processing times, typically 15 samples in less than 25 minutes.
  • Ensures objective data output for both quantitative and purity measurements.
  • For research use only; not for use in diagnostic procedures.
Catalog No. NC1940388 Supplier Agilent Technologies Supplier No. 50675588
The D5000 ScreenTape is designed for the automated electrophoresis of DNA samples, facilitating objective quantification, sizing, and purity determination. This system is essential for quality control in next-generation sequencing (NGS) workflows, as well as for the analysis of PCR and qPCR products. Compatible with all TapeStation instruments, the system utilizes individual lanes and loading tips to prevent cross-contamination and sample carryover.
  • Provides reliable analysis of DNA fragments and smears from 100 bp to 5000 bp.
  • Includes 7 ScreenTape devices, providing a total capacity of 105 samples.
  • Requires a 1 μL sample volume per analysis.
  • Features rapid processing times, typically 15 samples in less than 25 minutes.
  • Ensures objective data output for both quantitative and purity measurements.
  • For research use only; not for use in diagnostic procedures.