Learn More
ProSci D55 Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 89882 |
| Accession Number | NP_277051.3, NP_001001874.1, NP_001001875.1 |
| Common Name | tumor protein D52-like 3 |
| Content And Storage | Store at -20°C. When reconstituted minimize freezing and thawing. |
| Format | Lyophilized |
| Formulation | D55 peptide is supplied as 100 μg of dried peptide. When the peptide is reconstituted in 200 μL water, the concentration will be 0.5 mg/mL |
| For Use With (Application) | Blocking Assay |
| Gene Symbol | TPD52L3 |
| Quantity | 0.1 mg |
| Sequence | D55 peptide corresponds to 17 amino acids of D55. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.