Learn More
Medchemexpress LLC D5D-IN-326 | 1236767-85-3 | 99.9% | 473.27 | C17H11F8N3O4 | 10 MG
SDP
Catalog No. 5000227097
Encompass_Preferred
D5D-IN-326 is a selective, orally active delta-5 desaturase (D5D) inhibitor used in preclinical research. It demonstrates low-nanomolar potency in enzymatic and cell-based assays and has been reported to improve metabolic parameters in diet-induced obese mice. Documentation typically includes a product datasheet, certificate of analysis, and safety data sheet.
  • Selective delta-5 desaturase inhibition with low-nanomolar potency.
  • Demonstrated activity in enzymatic and cell-based assays.
  • Reduces insulin resistance and body weight in preclinical mouse models.
  • High purity (99.9% by LC-MS) confirmed by certificate of analysis.
  • Supplied as a small-molecule research reagent with supporting documentation for reproducibility.
Catalog No. 50-002-27097 Supplier Medchemexpress LLC Supplier No. HY11742710MG
D5D-IN-326 is a selective, orally active delta-5 desaturase (D5D) inhibitor used in preclinical research. It demonstrates low-nanomolar potency in enzymatic and cell-based assays and has been reported to improve metabolic parameters in diet-induced obese mice. Documentation typically includes a product datasheet, certificate of analysis, and safety data sheet.
  • Selective delta-5 desaturase inhibition with low-nanomolar potency.
  • Demonstrated activity in enzymatic and cell-based assays.
  • Reduces insulin resistance and body weight in preclinical mouse models.
  • High purity (99.9% by LC-MS) confirmed by certificate of analysis.
  • Supplied as a small-molecule research reagent with supporting documentation for reproducibility.