Learn More
Medchemexpress LLC D5D-IN-326 | 1236767-85-3 | 99.9% | 473.27 | C17H11F8N3O4 | 5 MG
SDP
Catalog No. 5000228106
Encompass_Preferred
D5D-IN-326 is a selective, orally active delta-5 desaturase (D5D) inhibitor (CAS 1236767-85-3) with reported nanomolar potency in enzymatic and cell-based assays and demonstrated activity in preclinical metabolic disease models. The compound is supplied for research use in multiple formats and sizes and is characterized by high purity and defined storage stability.
  • Selective inhibitor of delta-5 desaturase with nanomolar potency.
  • Reduces insulin resistance and body weight in preclinical models.
  • High purity (99.9%) for reliable experimental results.
  • Available as powder and solution formats for flexible dosing.
  • Stable under recommended storage conditions for long-term use.
  • Supplied in small research quantities suitable for in vitro and in vivo studies.
Catalog No. 50-002-28106 Supplier Medchemexpress LLC Supplier No. HY1174275MG
D5D-IN-326 is a selective, orally active delta-5 desaturase (D5D) inhibitor (CAS 1236767-85-3) with reported nanomolar potency in enzymatic and cell-based assays and demonstrated activity in preclinical metabolic disease models. The compound is supplied for research use in multiple formats and sizes and is characterized by high purity and defined storage stability.
  • Selective inhibitor of delta-5 desaturase with nanomolar potency.
  • Reduces insulin resistance and body weight in preclinical models.
  • High purity (99.9%) for reliable experimental results.
  • Available as powder and solution formats for flexible dosing.
  • Stable under recommended storage conditions for long-term use.
  • Supplied in small research quantities suitable for in vitro and in vivo studies.