Learn More
Grainger D6073 Inventory Control Label 2 in H PK1000
Catalog No. 502766079
Encompass_Preferred
The TapeCase Inventory Control Label is a 2" x 1" white label designed for inventory control and identification. It features a write-on surface and a permanent acrylic adhesive for application on bins, shelves, drawers, and more. The label's adhesive bonds instantly to provide a long-lasting label solution. This pack from TapeCase contains 500 labels per roll for efficient inventory management. With a temperature resistance range of -40°F to 200°F, these labels are suitable for indoor and outdoor use.
Catalog No. 50-276-6079 Supplier Grainger Supplier No. 16U827
The TapeCase Inventory Control Label is a 2" x 1" white label designed for inventory control and identification. It features a write-on surface and a permanent acrylic adhesive for application on bins, shelves, drawers, and more. The label's adhesive bonds instantly to provide a long-lasting label solution. This pack from TapeCase contains 500 labels per roll for efficient inventory management. With a temperature resistance range of -40°F to 200°F, these labels are suitable for indoor and outdoor use.