Learn More
Grainger D6073 Inventory Control Label Red PK1000
Catalog No. 502768412
Encompass_Preferred
The Grainger Approved Inventory Control Label is a white, adhesive label designed for inventory control and tracking applications. It measures 4 inches wide by 6 inches tall and features 30 labels per roll. The label has a write-on surface that accepts pen, marker, pencil, crayon and more. It adheres to paper, cardboard, plastic, metal and more. The adhesive removes cleanly after use. This versatile label provides an organized solution for warehouse, maintenance, facilities and office inventory tasks.
Catalog No. 50-276-8412 Supplier Grainger Supplier No. 16U945
The Grainger Approved Inventory Control Label is a white, adhesive label designed for inventory control and tracking applications. It measures 4 inches wide by 6 inches tall and features 30 labels per roll. The label has a write-on surface that accepts pen, marker, pencil, crayon and more. It adheres to paper, cardboard, plastic, metal and more. The adhesive removes cleanly after use. This versatile label provides an organized solution for warehouse, maintenance, facilities and office inventory tasks.