Learn More
Grainger D6074 Inventory Control Label Orange PK1000
Catalog No. 502768321
Encompass_Preferred
The Grainger 16U929 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning and scouring. It features an open web construction with non-scratching fibers that are ideal for cleaning pots, pans, dishes, countertops, sinks, showers, tubs, fiberglass, porcelain, stainless steel and other surfaces. This green hand pad measures 6 inches by 9 inches and has a medium duty abrasiveness. It is durable, rinses easily and is latex-free. The 3M Scotch-Brite General Purpose Hand Pad can tackle tough baked-on spills and stains without damaging surfaces.
Catalog No. 50-276-8321 Supplier Grainger Supplier No. 16U929
The Grainger 16U929 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning and scouring. It features an open web construction with non-scratching fibers that are ideal for cleaning pots, pans, dishes, countertops, sinks, showers, tubs, fiberglass, porcelain, stainless steel and other surfaces. This green hand pad measures 6 inches by 9 inches and has a medium duty abrasiveness. It is durable, rinses easily and is latex-free. The 3M Scotch-Brite General Purpose Hand Pad can tackle tough baked-on spills and stains without damaging surfaces.