Learn More
Grainger D6076 Inventory Control Label Red PK1000
Catalog No. 502765386
Encompass_Preferred
The Grainger 16U825 is a heavy duty steel shelving unit that provides durable storage space. This 4 shelf model has dimensions of 36" W x 18" D x 72" H and can hold up to 800 lbs per shelf. It features a boltless design for easy assembly and adjustable shelves that can be repositioned in 1-1/2" increments. The shelves are constructed from steel with a black powder coated finish. This shelving unit is ideal for warehouses, storage rooms, garages, and other industrial settings that require sturdy shelving.
Catalog No. 50-276-5386 Supplier Grainger Supplier No. 16U825
The Grainger 16U825 is a heavy duty steel shelving unit that provides durable storage space. This 4 shelf model has dimensions of 36" W x 18" D x 72" H and can hold up to 800 lbs per shelf. It features a boltless design for easy assembly and adjustable shelves that can be repositioned in 1-1/2" increments. The shelves are constructed from steel with a black powder coated finish. This shelving unit is ideal for warehouses, storage rooms, garages, and other industrial settings that require sturdy shelving.