Learn More
Enzo Life Sciences D609 (5 mg). CAS: 83373-60-8
SDP
Catalog No. 502012160
Encompass_Preferred
Selective inhibitor of phosphatidylcholine-specific phospholipase C (PC-PLC). Has been used to study the coupling of phosphatidylcholine-specific phospholipase C with sphingomyelinase. Shows antitumor and antiviral activity. Inhibits basement membrane collagen synthesis, which is the last step in the development of a new blood vessel. Inhibits induction of nitric oxide synthases (NOS). Induces apoptosis. It indirectly inhibits phorbol ester stimulated PLD activity. Alternative name: Tricyclodecan-9-yl xanthogenate . K. Purity: ≥98% (TLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2160 Supplier Enzo Life Sciences Supplier No. BMLST3300005
Selective inhibitor of phosphatidylcholine-specific phospholipase C (PC-PLC). Has been used to study the coupling of phosphatidylcholine-specific phospholipase C with sphingomyelinase. Shows antitumor and antiviral activity. Inhibits basement membrane collagen synthesis, which is the last step in the development of a new blood vessel. Inhibits induction of nitric oxide synthases (NOS). Induces apoptosis. It indirectly inhibits phorbol ester stimulated PLD activity. Alternative name: Tricyclodecan-9-yl xanthogenate . K. Purity: ≥98% (TLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.