Learn More
Cayman Chemical D609 (potassium salt), Purity: ≥90%, CAS: 83373-60-8, 10 mg
SDP
Catalog No. 502951886
Encompass_Preferred
A competitive inhibitor of PC-PLC (Ki = 6.4 µM); has no effect on bacterial phosphatidylinositol-specific PLC, bovine pancreatic PLA2, or phospholipase D; reduces sphingomyelin synthase activity by 55.5 and 90.5% in membrane preparations when used at concentrations of 100 and 200 mg/ml, respectively; has antioxidant, antiviral, and anti-tumor activities; inhibits LPS-stimulated NOS expression in phagocytes (IC50 = 20 µg/ml) and IL-1β-induced VCAM-1 expression in endothelial cells
Catalog No. 50-295-1886 Supplier Cayman Chemical Supplier No. 1330710 MG
A competitive inhibitor of PC-PLC (Ki = 6.4 µM); has no effect on bacterial phosphatidylinositol-specific PLC, bovine pancreatic PLA2, or phospholipase D; reduces sphingomyelin synthase activity by 55.5 and 90.5% in membrane preparations when used at concentrations of 100 and 200 mg/ml, respectively; has antioxidant, antiviral, and anti-tumor activities; inhibits LPS-stimulated NOS expression in phagocytes (IC50 = 20 µg/ml) and IL-1β-induced VCAM-1 expression in endothelial cells