Learn More
Biolegend DyLight™ 649 Donkey anti-rabbit IgG (min. x-reactivity)
Catalog No. 50169146
Encompass_Preferred
DyLight™ 649 Donkey anti-rabbit IgG (min. x-reactivity) [Poly4064]; Isotype: Donkey Polyclonal Ig; Reactivity: Rabbit; Apps: ICC, FC; Size: 100 ug
Catalog No. 50-169-146 Supplier Biolegend Supplier No. 406406
DyLight™ 649 Donkey anti-rabbit IgG (min. x-reactivity) [Poly4064]; Isotype: Donkey Polyclonal Ig; Reactivity: Rabbit; Apps: ICC, FC; Size: 100 ug