Learn More
TARGETMOL CHEMICALS INC D75-4590 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6119410MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. D75-4590 is a (beta)-16-Glucan inhibitor. As glucan is a major component of the cell wall specifically (beta)-16-Glucan D75-4590 can fight fungal infections by inhibiting its synthetase. purity: 99%
Catalog No. 50-257-9540