Learn More
Grainger Grainger Inc Outdoor Storage Shed, Steel, 248 cu. ft. 1 ea
Catalog No. 502771745
Encompass_Preferred
This outdoor storage shed provides a secure, durable solution for exterior storage needs. Engineered with steel construction and a 31 ga. frame, the unit is designed for long-lasting use in various outdoor environments. It features a spacious interior and a single hinged door for accessible entry. The unit is supplied unassembled and requires manual construction via a bolted assembly method.
  • Durable steel wall and roof construction
  • Single hinged door design with included latch
  • Slope roof design to facilitate drainage
  • Lockable design for enhanced security
  • Bolted assembly requires user installation
Catalog No. 50-277-1745 Supplier Grainger Supplier No. 13X110
This outdoor storage shed provides a secure, durable solution for exterior storage needs. Engineered with steel construction and a 31 ga. frame, the unit is designed for long-lasting use in various outdoor environments. It features a spacious interior and a single hinged door for accessible entry. The unit is supplied unassembled and requires manual construction via a bolted assembly method.
  • Durable steel wall and roof construction
  • Single hinged door design with included latch
  • Slope roof design to facilitate drainage
  • Lockable design for enhanced security
  • Bolted assembly requires user installation