Learn More
Grainger Ansell Microflex Nitrile Disposable Gloves, Cobalt Blue, Powder-Free, 100/Pk.
Catalog No. 502772377
Encompass_Preferred
These durable cobalt blue nitrile disposable gloves provide reliable hand protection and a secure, fully textured grip in both wet and dry conditions. Designed for versatility, they are well-suited for general-purpose tasks including light-duty cleaning, maintenance, and food preparation.
  • Ambidextrous design for easy donning
  • Beaded cuff for added strength and easy removal
  • Full texture provides enhanced gripping capabilities
  • Suitable for food processing and contact applications
  • Powder-free and not made from natural rubber latex to prevent Type I latex allergies
  • Complies with FDA 21 CFR 177.2600 for food contact
  • Manufactured under ISO 9001 certified quality systems
  • Palm thickness of 0.10 mm and finger thickness of 0.11 mm
Catalog No. 50-277-2377 Supplier Grainger Supplier No. 13U763
These durable cobalt blue nitrile disposable gloves provide reliable hand protection and a secure, fully textured grip in both wet and dry conditions. Designed for versatility, they are well-suited for general-purpose tasks including light-duty cleaning, maintenance, and food preparation.
  • Ambidextrous design for easy donning
  • Beaded cuff for added strength and easy removal
  • Full texture provides enhanced gripping capabilities
  • Suitable for food processing and contact applications
  • Powder-free and not made from natural rubber latex to prevent Type I latex allergies
  • Complies with FDA 21 CFR 177.2600 for food contact
  • Manufactured under ISO 9001 certified quality systems
  • Palm thickness of 0.10 mm and finger thickness of 0.11 mm