Learn More
Grainger Ansell Microflex N27 Nitrile Gloves, Cobalt Blue, Large, 100/Case
Catalog No. 502772651
Encompass_Preferred
These cobalt blue nitrile gloves are designed for high visibility and enhanced grip in multi-purpose applications. Featuring a fully textured surface, they provide secure handling in both wet and dry conditions, making them suitable for tasks ranging from life sciences and food processing to maintenance and light-duty cleaning.
  • Provides visual contrast for enhanced workplace safety
  • Fully textured surface ensures excellent grip
  • Powder-free construction with beaded cuff
  • Not made from natural rubber latex to prevent Type I allergies
  • Approved for food contact per FDA 21 CFR 177-2600
  • Compliant with EN ISO 21420:2020 standards
  • Palm thickness: 0.10 mm; finger thickness: 0.11 mm
Catalog No. 50-277-2651 Supplier Grainger Supplier No. 13U765
These cobalt blue nitrile gloves are designed for high visibility and enhanced grip in multi-purpose applications. Featuring a fully textured surface, they provide secure handling in both wet and dry conditions, making them suitable for tasks ranging from life sciences and food processing to maintenance and light-duty cleaning.
  • Provides visual contrast for enhanced workplace safety
  • Fully textured surface ensures excellent grip
  • Powder-free construction with beaded cuff
  • Not made from natural rubber latex to prevent Type I allergies
  • Approved for food contact per FDA 21 CFR 177-2600
  • Compliant with EN ISO 21420:2020 standards
  • Palm thickness: 0.10 mm; finger thickness: 0.11 mm