Learn More
Grainger Brady® MiniMark™ Printer Ribbon, Resin, Black, 4.4 in., 2/Pk.
Catalog No. 502794517
Encompass_Preferred
MiniMark™ Series printer ribbons are pre-loaded onto spindles with keyed end caps for fast, hassle-free installation. This one-color ribbon is designed for printing a single color onto labels and is formulated with the R7968 resin series for reliable performance.
  • Provides standard scratch, smear, and chemical resistance when printed on compatible materials.
  • Compatible with B-7569 adhesive vinyl.
  • Features keyed spindles to ensure the ribbon always faces the correct direction during use.
  • Ribbon dimensions measure 111.76 mm (4.4 in.) in width and 88.4 m (290 ft.) in length.
  • Supplied in a pack of two rolls.
Catalog No. 50-279-4517 Supplier Grainger Supplier No. 1AMV4
MiniMark™ Series printer ribbons are pre-loaded onto spindles with keyed end caps for fast, hassle-free installation. This one-color ribbon is designed for printing a single color onto labels and is formulated with the R7968 resin series for reliable performance.
  • Provides standard scratch, smear, and chemical resistance when printed on compatible materials.
  • Compatible with B-7569 adhesive vinyl.
  • Features keyed spindles to ensure the ribbon always faces the correct direction during use.
  • Ribbon dimensions measure 111.76 mm (4.4 in.) in width and 88.4 m (290 ft.) in length.
  • Supplied in a pack of two rolls.