Learn More
Grainger Brady® MiniMark Series Printer Ribbon, Resin, Red, 2/Pk.
Catalog No. 502795065
Encompass_Preferred
These single-color printer ribbons are designed for use with the MiniMark industrial label printer. Each ribbon is pre-loaded onto a spindle featuring keyed end caps to ensure correct orientation and facilitate fast, hassle-free installation. The R7968 series resin formulation provides reliable scratch, smear, and chemical resistance when printed on compatible label materials, such as B-7569 adhesive vinyl.
  • Designed for single-color printing applications
  • Features R7968 series resin formulation for durability
  • Compatible with B-7569 adhesive vinyl
  • Spindles ensure ribbon always faces in the right direction
  • Supplied pre-loaded on keyed spindles for easy installation
Catalog No. 50-279-5065 Supplier Grainger Supplier No. 1AMV6
These single-color printer ribbons are designed for use with the MiniMark industrial label printer. Each ribbon is pre-loaded onto a spindle featuring keyed end caps to ensure correct orientation and facilitate fast, hassle-free installation. The R7968 series resin formulation provides reliable scratch, smear, and chemical resistance when printed on compatible label materials, such as B-7569 adhesive vinyl.
  • Designed for single-color printing applications
  • Features R7968 series resin formulation for durability
  • Compatible with B-7569 adhesive vinyl
  • Spindles ensure ribbon always faces in the right direction
  • Supplied pre-loaded on keyed spindles for easy installation