Learn More
Grainger Brady Single-Color Printer Ribbon, Resin, Green, 4 7/16 in. x 290 ft., 2/Pk.
Catalog No. 502794518
Encompass_Preferred
These green, single-color resin printer ribbons are designed for use with the MiniMark™ industrial label maker. The ribbons are pre-loaded into cartridges to eliminate the need for installing separate ink or printing fluids, providing a streamlined labeling solution for various facility identification tasks.
  • Features R7968 series resin formulation for scratch, smear, and chemical resistance.
  • Includes spindles with keyed end caps for rapid, error-free installation.
  • Optimized for printing on B-7569 adhesive vinyl materials.
  • Ribbon dimensions measure 11.2 cm (4.4 in.) in width by 88.4 m (290 ft.) in length.
  • Compliant with RoHS 2002/95/EC and 2005/618/EC standards.
Catalog No. 50-279-4518 Supplier Grainger Supplier No. 1AMV8
These green, single-color resin printer ribbons are designed for use with the MiniMark™ industrial label maker. The ribbons are pre-loaded into cartridges to eliminate the need for installing separate ink or printing fluids, providing a streamlined labeling solution for various facility identification tasks.
  • Features R7968 series resin formulation for scratch, smear, and chemical resistance.
  • Includes spindles with keyed end caps for rapid, error-free installation.
  • Optimized for printing on B-7569 adhesive vinyl materials.
  • Ribbon dimensions measure 11.2 cm (4.4 in.) in width by 88.4 m (290 ft.) in length.
  • Compliant with RoHS 2002/95/EC and 2005/618/EC standards.