Learn More
Aviva Systems Biology Corp D9Ertd402e Antibody - N-terminal region (ARP54464_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54464_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against D9Ertd402e. It was validated on Western Blot. . Alternate names: Gm1129, MGC91156, TOAG-1, D9Ertd402eApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-929