Learn More
Med Vet International DA Toggle Suture, J0191, 2 Pack
Catalog No. 5000583166
Encompass_Preferred
Used in the Grymer/Sterner method for nonsurgical percutaneous fixation of the left displaced abomasum (LDA).
Catalog No. 50-005-83166 Supplier Med Vet International Supplier No. JOR191
Used in the Grymer/Sterner method for nonsurgical percutaneous fixation of the left displaced abomasum (LDA).