Learn More
Biocare Medical, LLC Da Vinci Green Diluent - 100 ml
SDP
Catalog No. 5082974
Encompass_Preferred
Da Vinci Green antibody diluent is a modified PBS formulation with protein carrier and preservative. This modified PBS is formulated for superior performance and stability. In addition, it is protease and IgG-free. Da Vinci Green is Biocare's new standard-universal PBS diluent. It can also be used on an automated staining system.
This item is not returnable. View return policy
Catalog No. 50-829-74 Supplier Biocare Medical, LLC Supplier No. PD900L
Da Vinci Green antibody diluent is a modified PBS formulation with protein carrier and preservative. This modified PBS is formulated for superior performance and stability. In addition, it is protease and IgG-free. Da Vinci Green is Biocare's new standard-universal PBS diluent. It can also be used on an automated staining system.