Learn More
Medchemexpress LLC DAA-1106 | 220551-92-8 | 99.9% | 395.42 | 100 MG
SDP
Catalog No. 5000401919
Encompass_Preferred
DAA1106 is a peripheral benzodiazepine receptor (PBR) agonist that is permeable to the blood-brain barrier, but has no affinity for GABAA receptors.
  • Acts as a peripheral benzodiazepine receptor agonist
  • Permeable to the blood-brain barrier
  • No affinity for GABAA receptors
Catalog No. 50-004-01919 Supplier Medchemexpress LLC Supplier No. HY19945100MG
DAA1106 is a peripheral benzodiazepine receptor (PBR) agonist that is permeable to the blood-brain barrier, but has no affinity for GABAA receptors.
  • Acts as a peripheral benzodiazepine receptor agonist
  • Permeable to the blood-brain barrier
  • No affinity for GABAA receptors