Learn More
AdipoGen DAABD-AE
SDP
Supplier:  AdipoGen CDXD0204M050
Encompass_Preferred
Chemical. CAS 913253-56-2. Formula C12H20N6O3S. MW 328.39. Synthetic. 4-2-N,N-dimethylaminoethylaminosulfonyl-7-2-aminoethylamino-2,1,3-benzoxadiazole DAABD-AE is new derivatization reagent for rapid and specific liquid chromatography tandem mass spectrometric quantification of pristanic acid, phytanic acid, and very long chain fatty acids i.e. hexacosanoic, tetracosanoic, and docosanoic acids in plasma.
Catalog No. 50-256-1188