Learn More
Abcam DAABD-AE, Derivatization reagent, 50MG
Catalog No. 502705326
Encompass_Preferred
MW 328.39 g/mol, Purity >95%. Derivatization reagent for rapid and specific liquid chromatography tandem mass spectrometric quantification of pristanic acid, phytanic acid, and very long chain fatty acids in plasma.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5326 Supplier Abcam Supplier No. AB14529050MG
MW 328.39 g/mol, Purity >95%. Derivatization reagent for rapid and specific liquid chromatography tandem mass spectrometric quantification of pristanic acid, phytanic acid, and very long chain fatty acids in plasma.
The product is subject to the following: Abcam Restricted Use Statement