Learn More
Medchemexpress LLC HY-111412 10mg Medchemexpress, DAAO inhibitor-1 CAS:2065250-25-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11141210MG
Encompass_Preferred
Medchemexpress, HY-111412 10mg DAAO inhibitor-1 CAS:2065250-25-9 DAAO inhibitor-1 is a potent D-amino acid oxidase (DAAO) inhibitor with an IC50 of 0.12 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8860