Learn More
Vector Laboratories Dolichos Biflorus Agglutinin (DBA) Rhodamine-labeled 2 mg
SDP
Supplier:  Vector Laboratories RL10322
Encompass
Dolichos biflorus agglutinin (DBA) binds α-linked N-acetylgalactosamine. DBA is a glycoprotein consisting of four subunits of approximately equal size with a molecular weight of about 111 kDa and is used to establish secretor status in blood group A individuals by hemagglutination inhibition techniques and for blood typing. It has also been used as a general marker of developing renal collecting ducts. This rhodamine-labeled DBA features a ratio of fluorophore to lectin protein that provides optimal staining (excitation 575 nm, emission 600 nm) and is supplied in solution essentially free of unconjugated fluorophore.
Catalog No. NC0127925