Learn More
Diagnostic Biosystems DAB AUTO DAB Auto
Supplier:  Diagnostic Biosystems K081AUTO
Encompass_Preferred
DAB, a widely used chromogen for immunoperoxidase staining, is well accepted among pathologists because of its increased sensitivity and ability to give cleaner background as compared to amino ethylcarbazole (AEC). Specimens stained in DAB can be dehydrated, cleared, and mounted for permanent record keeping.
Catalog No. 50-240-7597