Learn More
Seracare Life Sciences Inc DAB-C SUBSTRATE CONCENTRATE
Catalog No. 5067563
Encompass_Preferred
Available as a substrate component of the HistoMark Orange immunohistochemistry substrate kit or as a stand-alone product. Requires Enhance Orange Buffer and Peroxide Solution for use.
This item is not returnable. View return policy
Catalog No. 50-675-63 Supplier Seracare Life Sciences Inc Supplier No. 55100043
Available as a substrate component of the HistoMark Orange immunohistochemistry substrate kit or as a stand-alone product. Requires Enhance Orange Buffer and Peroxide Solution for use.