Learn More
Biocare Medical, LLC DAB Chromogen Kit - 100 ml
SDP
Catalog No. 5082814
Encompass_Preferred
3,3’-Diaminobenzidine (DAB) is a widely used chromogen for immunohistochemical staining and immunoblotting. When in the presence of peroxidase enzyme, DAB produces a brown precipitate that is insoluble in alcohol and xylene. This product comes in a two-component system consisting of a liquid stable DAB chromogen and DAB substrate buffer. The color of the chromogen solution can vary from colorless to pale brown.
Catalog No. 50-828-14 Supplier Biocare Medical, LLC Supplier No. DB801L
3,3’-Diaminobenzidine (DAB) is a widely used chromogen for immunohistochemical staining and immunoblotting. When in the presence of peroxidase enzyme, DAB produces a brown precipitate that is insoluble in alcohol and xylene. This product comes in a two-component system consisting of a liquid stable DAB chromogen and DAB substrate buffer. The color of the chromogen solution can vary from colorless to pale brown.