Learn More
Electron Microscopy Sciences DAB-HCl 3,3-Diaminobenzidine Tetra Hydrochloride 1 GR

Supplier: Electron Microscopy Sciences 13080
CAS #7411-49-6 C₁₂H₁₄N₄·4HCl F.W. 360.11
Melting point: 300°C
Used for histochemical localization of peroxidase with DAB and in immunocytochemistry.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.