Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ DAB Quanto Detection System

Catalog No. p-4532346 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 L
125 mL DAB Quanto Substrate, 4 mL DAB Quanto Chromagen
60 mL DAB Quanto Substrate, 2 mL DAB Quanto Chromagen
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TA060QHDX 60 mL DAB Quanto Substrate, 2 mL DAB Quanto Chromagen
TA125QHDX 125 mL DAB Quanto Substrate, 4 mL DAB Quanto Chromagen
TA999QHDX 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
DAB Quanto Chromogen contains Diaminobenzidine (DAB) in buffer. DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Reagent is acidic and can cause burns if skin contact occurs. Handle with care and dispose of according to regulations. DAB Quanto Substrate contains Hydrogen Peroxide in buffer. Avoid contact with skin and eyes.
Catalog No. TA060QHDX Supplier Epredia™ Supplier No. TA060QHDX
Only null left
Add to Cart
Add to Cart
DAB Quanto Chromogen contains Diaminobenzidine (DAB) in buffer. DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Reagent is acidic and can cause burns if skin contact occurs. Handle with care and dispose of according to regulations. DAB Quanto Substrate contains Hydrogen Peroxide in buffer. Avoid contact with skin and eyes.

Provides a strong brown staining and can be used after reconstitution for up to 1 week at room temperature and up to 2 weeks at 2° to 8°C.

DAB Quanto has been validated in combination with both UltraVision LP and UltraVision Quanto and provides four major benefits.
    • Strong staining—ideal to quantifying cells and protein expression
    • Less mixing—reducing personnel time and stress
    • Less disposal—cuts down waste
    • Lower risk of mixing errors—supports higher staining consistency
    3,3' Diaminobenzidine (DAB) is a widely used chromogen for immunohistochemical staining and immunoblotting. The DAB Quanto is an entirely new formula for a diaminobenzidine-based peroxidase substrate that develops faster on tissues sections reducing incubation times. It utilizes novel chemistry to provide greater sensitivity, generating a crisper, darker, insoluble precipitate. The standard working dilution is 30μL of DAB Quanto Chromogen per 1ml of DAB Quanto Substrate. The use of liquid components reduces some risks associated with the handling of powders or tablets. The DAB Quanto offers an added advantage, as the working solution is stable for up to 2 weeks at 2 to 8°C and up to a week at room temperature enabling continued use and minimizing waste disposal.
    • Supplied in a two-component form consisting of a DAB Quanto Chromogen and DAB Quanto Substrate
    • Standard working dilution is 30μL of DAB Quanto Chromogen per 1mL of DAB Quanto Substrate
    • Use of liquid components reduces some risks associated with the handling of powders or tablets
  • Specifications

    Chromagen Type DAB Quanto Chromagen
    Content And Storage 2° to 8°C
    Quantity 60 mL DAB Quanto Substrate, 2 mL DAB Quanto Chromagen
    Substrate Type Dab Quanto Subrate
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.