Learn More
Seracare Life Sciences Inc DAB REAGENT SET
Catalog No. 5067498
Encompass_Preferred
This kit provides the most commonly used amine substrate for the histochemcial localization of peroxidases. DAB is an extremely sensitive indicator, producing a characteristic brown oxidation product easily visible with the light microscope. HistoMark DAB Reagent Set produces a sharply localized, brown deposit at the site of the reaction. Kit includes DAB Substrate Concentrate, Buffer Concentrate, and Peroxidase Solution.
This item is not returnable. View return policy
Catalog No. 50-674-98 Supplier Seracare Life Sciences Inc Supplier No. 55100031
This kit provides the most commonly used amine substrate for the histochemcial localization of peroxidases. DAB is an extremely sensitive indicator, producing a characteristic brown oxidation product easily visible with the light microscope. HistoMark DAB Reagent Set produces a sharply localized, brown deposit at the site of the reaction. Kit includes DAB Substrate Concentrate, Buffer Concentrate, and Peroxidase Solution.