Learn More
Biocare Medical, LLC DAB Sparkle - 25 ml
SDP
Catalog No. 5082822
Encompass_Preferred
This specially formulated ready-to-use enhancing solution will increase sensitivity and contrast of DAB stained tissues.
Catalog No. 50-828-22 Supplier Biocare Medical, LLC Supplier No. DS830H
This specially formulated ready-to-use enhancing solution will increase sensitivity and contrast of DAB stained tissues.