Learn More
Abcam DAB Substrate Kit, 125ML
Catalog No. NC0501765
Encompass_Preferred
DAB substrate kit is used for immunohistochemistry (IHC) staining using HRP/peroxidase enzyme for detection. - Cited in over 375 publications - Stable for up to 6 hours after mixing - Produces crisp brown stain, alcohol-insoluble - Convenient two-component liquid format - Refrigerator-stable for extended usability
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0501765 Supplier Abcam Supplier No. AB64238125ML
DAB substrate kit is used for immunohistochemistry (IHC) staining using HRP/peroxidase enzyme for detection. - Cited in over 375 publications - Stable for up to 6 hours after mixing - Produces crisp brown stain, alcohol-insoluble - Convenient two-component liquid format - Refrigerator-stable for extended usability
The product is subject to the following: Abcam Restricted Use Statement