Learn More
Seracare Life Sciences Inc DAB SUBSTRATE SOLUTION
Supplier:  Seracare Life Sciences Inc 55100041
Encompass_Preferred
DAB is the most commonly used amine substrate for the histochemcial localization of peroxidases. DAB is an extremely sensitive indicator, producing a characteristic brown oxidation product easily visible with light microscopy. DAB can be used as a stand alone product, or it may be combined with KPL's Enhance Black Buffer to create HistoMark BLACK substrate.
Catalog No. 50-675-58