Learn More
Aviva Systems Biology Corp DAB1 Antibody - middle region (ARP52396_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52396_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DAB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DAB1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53972