Learn More
Medchemexpress LLC DABCO-bis(sulfur dioxide) | 119752-83-9 | MFCD22377802 | 97.0% | 240.3 g/mol | C6H12N2O4S2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW03304950G
Encompass_Preferred
DABCO-bis(sulfur dioxide) (DABSO) is a bench-stable, crystalline reagent that serves as a convenient solid source of sulfur dioxide for organic synthesis. It is commonly used to introduce sulfonyl functionality and to prepare sulfonamides and sulfones without handling gaseous SO2.
  • Bench-stable solid source of sulfur dioxide.
  • Enables preparation of sulfonamides and sulfones.
  • Eliminates the need to handle gaseous sulfur dioxide.
  • High purity suitable for synthetic applications (97.0%).
  • Crystalline solid simplifies storage and handling.
Catalog No. 50-003-00085