Learn More
Medchemexpress LLC HY-D1045 100mg Medchemexpress, Dabcyl acid CAS:6268-49-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD1045100MG
Encompass_Preferred
Medchemexpress, HY-D1045 100mg Dabcyl acid CAS:6268-49-1 Dabcyl acid (Dabcyl) is the original dark fluorescence quencher. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2751