Learn More
CPC Scientific DABCYL-Ile-Lys-Thr-Glu-Glu-Ile-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-EDANS (acetate salt) 0.5MG
SDP
Catalog No. 502108716
Encompass
ONE-LETTER SEQUENCE: Dabcyl-IKTEEISEVNLDAEF-EDANSMOLECULAR FORMULA: C103H146N22O32S1MOLECULAR WEIGHT:2236.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Alzheimer's Disease
Catalog No. 50-210-8716 Supplier CPC Scientific Supplier No. AMYD100A
ONE-LETTER SEQUENCE: Dabcyl-IKTEEISEVNLDAEF-EDANSMOLECULAR FORMULA: C103H146N22O32S1MOLECULAR WEIGHT:2236.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Alzheimer's Disease