Learn More
CPC Scientific DABCYL-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-EDANS (acetate salt) 0.5MG
SDP
Catalog No. 502108718
Encompass
SEQUENCE: DABCYL-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-EDANS (acetate salt)ONE-LETTER SEQUENCE: Dabcyl-SEVNLDAEF-EDANSMOLECULAR FORMULA: C71H91N15O21S1MOLECULAR WEIGHT: 1522.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [1802078-36-9]SYNONYMS: DABCYL-(Asn,Leu)-Amyloid /A4 Protein Precursor (667-675)-EDANS, DABCYL-(ASN670,LEU671)-APP770 (667-675)-EDANS, DABCYL-(ASN670,LEU671)-AMYLOID BETA/A4 PROTEIN PRECURSOR770 (667-675)-EDANS, DABCYL-SER-GLU-VAL-ASN-LEU-ASP-ALA-GLU-PHE-EDANS, DABCYL-Asn670,Leu671-Amyloid b/A4 Protein Precursor770: 667-675-EDANSRESEARCH AREA: Alzheimer's DiseaseREFERENCES:SKU(s): AMYD-101A, AMYD-101B
Catalog No. 50-210-8718 Supplier CPC Scientific Supplier No. AMYD101A
SEQUENCE: DABCYL-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-EDANS (acetate salt)ONE-LETTER SEQUENCE: Dabcyl-SEVNLDAEF-EDANSMOLECULAR FORMULA: C71H91N15O21S1MOLECULAR WEIGHT: 1522.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [1802078-36-9]SYNONYMS: DABCYL-(Asn,Leu)-Amyloid /A4 Protein Precursor (667-675)-EDANS, DABCYL-(ASN670,LEU671)-APP770 (667-675)-EDANS, DABCYL-(ASN670,LEU671)-AMYLOID BETA/A4 PROTEIN PRECURSOR770 (667-675)-EDANS, DABCYL-SER-GLU-VAL-ASN-LEU-ASP-ALA-GLU-PHE-EDANS, DABCYL-Asn670,Leu671-Amyloid b/A4 Protein Precursor770: 667-675-EDANSRESEARCH AREA: Alzheimer's DiseaseREFERENCES:SKU(s): AMYD-101A, AMYD-101B