Learn More
CPC Scientific DABCYL-Val-Asn-Leu-Asp-Ala-Glu-EDANS (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108720
Encompass
SEQUENCE: DABCYL-Val-Asn-Leu-Asp-Ala-Glu-EDANS (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dabcyl-VNLDAE-EDANSMOLECULAR FORMULA: C54H70N12O15S1MOLECULAR WEIGHT: 1159.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [1802078-37-0]SYNONYMS: DABCYL-[Asn670, Leu671]-Amyloid /A4 Precursor Protein 770 Fragment 669-674-EDANSRESEARCH AREA: Alzheimer's DiseaseREFERENCES:SKU(s): AMYD-102A, AMYD-102B
Catalog No. 50-210-8720 Supplier CPC Scientific Supplier No. AMYD102A
SEQUENCE: DABCYL-Val-Asn-Leu-Asp-Ala-Glu-EDANS (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dabcyl-VNLDAE-EDANSMOLECULAR FORMULA: C54H70N12O15S1MOLECULAR WEIGHT: 1159.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [1802078-37-0]SYNONYMS: DABCYL-[Asn670, Leu671]-Amyloid /A4 Precursor Protein 770 Fragment 669-674-EDANSRESEARCH AREA: Alzheimer's DiseaseREFERENCES:SKU(s): AMYD-102A, AMYD-102B