Learn More
Ubpbio A FRET-based fluorogenic substrate of Papain-like protease (PLPro) from SARS viruses, including SARS-CoV-2. PLPro also has deubiquitinase activity. 5MG
Catalog No. 502339410
Encompass_Preferred
Dabcyl–FRLKGGAPIKGV–Edans is an internally quenched fluorogenic substrate for the SARS-CoV or MERS-CoV family Papain-like protease (PLPro or PLP) that recognizes a LXGG consensus cleavage sites. Upon PLP-mediated cleavage between glycine and alanine, the resulting APIKGV-Edans fluorescence can be monitored at the excitation/emission wavelengths of 355/538 nm, respectively. In addition, PLP has deubiquitinating and deISG15ylation activities.
Catalog No. 50-233-9410 Supplier Ubpbio Supplier No. V10205
Dabcyl–FRLKGGAPIKGV–Edans is an internally quenched fluorogenic substrate for the SARS-CoV or MERS-CoV family Papain-like protease (PLPro or PLP) that recognizes a LXGG consensus cleavage sites. Upon PLP-mediated cleavage between glycine and alanine, the resulting APIKGV-Edans fluorescence can be monitored at the excitation/emission wavelengths of 355/538 nm, respectively. In addition, PLP has deubiquitinating and deISG15ylation activities.